Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11FIP5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220035 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220035 Supplier Novus Biologicals Supplier No. NBP220035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB11FIP5 Polyclonal specifically detects RAB11FIP5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RAB11FIP5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.75mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9BXF6
Gene Alias DKFZp434H018, Gaf-1, GAF1rab11-FIP5, Gamma-SNAP-associated factor 1, KIAA0857gaf-1, Phosphoprotein pp75, pp75, RAB11 family interacting protein 5 (class I), Rab11-FIP5, Rab11-interacting protein Rip11, RAB11RIP5, RIP11rab11 family-interacting protein 5
Gene Symbols RAB11FIP5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB11-FIP5. The exact sequence is proprietary.
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26056
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.