Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11FIP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP18185525UL
Description
RAB11FIP5 Polyclonal specifically detects RAB11FIP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
RAB11FIP5 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockout Validated | |
Q9BXF6 | |
RAB11FIP5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SPHPVKPLSAAPVEGSPDRKQSRSSLSIALSSGLEKLKTVTSGSIQPVTQAPQAGQMVDTKRLKDSAVLDQSAKYYHLTHDELISLLLQRERELSQRDEHVQEL | |
Affinity Purified | |
RUO | |
26056 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp434H018, Gaf-1, GAF1rab11-FIP5, Gamma-SNAP-associated factor 1, KIAA0857gaf-1, Phosphoprotein pp75, pp75, RAB11 family interacting protein 5 (class I), Rab11-FIP5, Rab11-interacting protein Rip11, RAB11RIP5, RIP11rab11 family-interacting protein 5 | |
Rabbit | |
70 kDa | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction