Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11FIP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | RAB11FIP5 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RAB11FIP5 Polyclonal specifically detects RAB11FIP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
RAB11FIP5 | |
Polyclonal | |
Rabbit | |
Human | |
Q9BXF6 | |
26056 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SPHPVKPLSAAPVEGSPDRKQSRSSLSIALSSGLEKLKTVTSGSIQPVTQAPQAGQMVDTKRLKDSAVLDQSAKYYHLTHDELISLLLQRERELSQRDEHVQEL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
70 kDa |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp434H018, Gaf-1, GAF1rab11-FIP5, Gamma-SNAP-associated factor 1, KIAA0857gaf-1, Phosphoprotein pp75, pp75, RAB11 family interacting protein 5 (class I), Rab11-FIP5, Rab11-interacting protein Rip11, RAB11RIP5, RIP11rab11 family-interacting protein 5 | |
RAB11FIP5 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title