Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP100132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP100132 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100132 Supplier Novus Biologicals Supplier No. NBP100132
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Rab17 Polyclonal specifically detects Rab17 in Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Rab17
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.2 - 0.6 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_033024.1
Gene Alias FLJ12538, RAB17, member RAS oncogene family, ras-related protein Rab-17
Gene Symbols RAB17
Host Species Goat
Immunogen Peptide with sequence C-RKDSFHKAQQWLED corresponding to internal region according to Uniprot Mouse NP_033024.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64284
Test Specificity Reported variants represent identical protein (NP_001153197.2; NP_033024.1).
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.