Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB17 Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene
SDP

Catalog No. 502542426
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2426 30 μL
50-167-2401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2426 Supplier OriGene Supplier No. TA501258S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB17 Monoclonal antibody specifically detects RAB17 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RAB17
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4E7
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene H6PD
Gene Accession No. O95479
Gene Alias AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478
Host Species Mouse
Immunogen Full length human recombinant protein of human H6PD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9563
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.