Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB18 Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB180861 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180861 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180861 Supplier Novus Biologicals Supplier No. NBP306453R
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAB18 Polyclonal antibody specifically detects RAB18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RAB18
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias RAB18 small GTPase, RAB18, member RAS oncogene family, RAB18LI1, ras-related protein Rab-18
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RAB18 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22931
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.