Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220038 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220038 Supplier Novus Biologicals Supplier No. NBP220038

Rabbit Polyclonal Antibody

Rab2 Polyclonal specifically detects Rab2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Rab2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.42mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P61019
Gene Alias RAB2A, member RAS oncogene family, RAB2RAB2, member RAS oncogene family, ras-related protein Rab-2A, small GTP binding protein RAB2A
Gene Symbols RAB2A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB2A. The exact sequence is proprietary.
Molecular Weight of Antigen 24 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5862
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Dog (100%), Rhesus Monkey (100%), Rabbit (100%), Zebrafish (100%), Bovine (100%), Chimpanzee (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.