Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RAB23 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RAB23 Polyclonal specifically detects RAB23 in Mouse samples. It is validated for Western Blot.Specifications
| RAB23 | |
| Polyclonal | |
| Rabbit | |
| NP_033025 | |
| 51715 | |
| Synthetic peptide directed towards the middle region of human Rab23The immunogen for this antibody is Rab23. Peptide sequence DPEQTHSSSNKIGVFNASVRSHLGQNSSSLNGGDVINLRPNKQRTKRTRN. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23 | |
| RAB23 | |
| IgG | |
| 26 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title