Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab24 Antibody (OTI3H1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N273770A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273770A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273770A700 Supplier Novus Biologicals Supplier No. NBP273770AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab24 Monoclonal specifically detects Rab24 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rab24
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3H1
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias RAB24, member RAS oncogene family, ras-related protein Rab-24
Gene Symbols RAB24
Host Species Mouse
Immunogen Full length recombinant protein of human Rab24 (NP_001026847) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 53917.0
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.