Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab25 Antibody (27H3), Novus Biologicals™
SDP

Catalog No. NB298269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298269 100 μL
NB298268 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298269 Supplier Novus Biologicals Supplier No. NBP326354100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rab25 Monoclonal antibody specifically detects Rab25 in Human samples. It is validated for ELISA, Immunocytochemistry

Specifications

Antigen Rab25
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 27H3
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Rab25 [UniProt P57735]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57111
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.