Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab25 Antibody (27H3), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $479.73
Specifications
| Antigen | Rab25 |
|---|---|
| Clone | 27H3 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
Description
Rab25 Monoclonal antibody specifically detects Rab25 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| Rab25 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 57111 | |
| IgG | |
| Affinity purified |
| 27H3 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CATX8, CATX-8RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
| A synthesized peptide derived from Human Rab25 [UniProt P57735] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title