Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179484
Description
RAB35 Polyclonal specifically detects RAB35 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RAB35 | |
Polyclonal | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
GTP-binding protein RAY, H-ray, RAB1C, RAB35, member RAS oncogene family, Ras-related protein Rab-1C, ras-related protein rab-1c (GTP-binding protein ray), ras-related protein Rab-35, RAY | |
Rabbit | |
23 kDa | |
100 μL | |
Primary | |
Zebrafish: 79%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
NP_006852 | |
RAB35 | |
Synthetic peptide directed towards the middle region of human RAB35. Peptide sequence: GIQLFETSAKENVNVEEMFNCITELVLRAKKDNLAKQQQQQQNDVVKLTK | |
Affinity purified | |
RUO | |
11021 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction