Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RAB35 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17948420
![]() |
Novus Biologicals
NBP17948420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179484
![]() |
Novus Biologicals
NBP179484 |
100 μL |
Each for $487.50
|
|
|||||
Description
RAB35 Polyclonal specifically detects RAB35 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RAB35 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
GTP-binding protein RAY, H-ray, RAB1C, RAB35, member RAS oncogene family, Ras-related protein Rab-1C, ras-related protein rab-1c (GTP-binding protein ray), ras-related protein Rab-35, RAY | |
RAB35 | |
IgG | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_006852 | |
11021 | |
Synthetic peptide directed towards the middle region of human RAB35. Peptide sequence: GIQLFETSAKENVNVEEMFNCITELVLRAKKDNLAKQQQQQQNDVVKLTK | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title