Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab3A Antibody (OTI1G3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP273773B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273773B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273773B Supplier Novus Biologicals Supplier No. NBP273773B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab3A Monoclonal specifically detects Rab3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Rab3A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G3
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias RAB3A, member RAS oncogene family, RAS-associated protein RAB3A, ras-related protein Rab-3A
Gene Symbols RAB3A
Host Species Mouse
Immunogen Full length recombinant protein of human Rab3A (NP_002857) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5864.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.