Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab3D Antibody (5E9), Novus Biologicals™
SDP

Catalog No. NB159000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB159000 Supplier Novus Biologicals Supplier No. NBP314764
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rab3D Monoclonal antibody specifically detects Rab3D in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Rab3D
Applications Western Blot
Classification Monoclonal
Clone 5000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.4, 50% glycerol
Gene Alias D2-2, GOVras-related protein Rab-3D, RAB16glioblastoma overexpressed, RAB3D, member RAS oncogene family, RAD3DRab3D upregulated with myeloid differentiation
Host Species Mouse
Immunogen Full length recombinant Rab3D protein (Accession #NP_004274)
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9545
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.