Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3GAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP119154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119154 Supplier Novus Biologicals Supplier No. NBP119154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB3GAP1 Polyclonal specifically detects RAB3GAP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen RAB3GAP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q15042
Gene Alias KIAA0066P130, RAB3 GTPase activating protein subunit 1 (catalytic), RAB3 GTPase-activating protein 130 kDa subunit, rab3 GTPase-activating protein catalytic subunit, Rab3-GAP, Rab3-GAP p130, RAB3GAPDKFZp434A012, WARBM1RAB3GAP130
Gene Symbols RAB3GAP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human RAB3 GTPase activating protein subunit 1 using the numbering given in entry NP_036365.1 (GeneID 22930).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22930
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.