Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3IL1 Antibody (OTI1B5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273774IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273774IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273774IR Supplier Novus Biologicals Supplier No. NBP273774IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB3IL1 Monoclonal specifically detects RAB3IL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen RAB3IL1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1B5
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GRAB, guanine nucleotide exchange factor for Rab3A, guanine nucleotide exchange factor for Rab-3A, RAB3A interacting protein (rabin3)-like 1, Rab-3A-interacting-like protein 1, Rab3A-interacting-like protein 1, Rabin3-like 1
Gene Symbols RAB3IL1
Host Species Mouse
Immunogen Full length recombinant protein of human RAB3IL1 (NP_037533) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5866.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.