Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3IP Antibody (OTI5F2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27377A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27377A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27377A30 Supplier Novus Biologicals Supplier No. NBP273775AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB3IP Monoclonal specifically detects RAB3IP in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RAB3IP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5F2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ14660, FLJ22548, MGC71495, RAB3A interacting protein (rabin3), rab-3A-interacting protein, Rab3A-interacting protein, rabin-3, RABIN3, SSX2 interacting protein, SSX2-interacting protein
Gene Symbols RAB3IP
Host Species Mouse
Immunogen Full length recombinant protein of human RAB3IP (NP_071901) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 117177.0
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.