Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB171854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171854 100 μg
NB171855 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171854 Supplier Novus Biologicals Supplier No. NBP318233100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rab4 Polyclonal antibody specifically detects Rab4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Rab4
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:150
Formulation PBS pH 7.4, 50% glycerol
Gene Alias Oncogene RAB4, RAB4, member RAS oncogene family, RAB4A, member RAS oncogene family, RAB4HRES-1/RAB4, ras-related protein Rab-4A
Host Species Rabbit
Immunogen C-terminal peptide from human Rab4
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5867
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.