Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB43 Mouse anti-Human, Clone: 3B2A8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | RAB43 |
|---|---|
| Clone | 3B2A8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7072
![]()
|
Proteintech Group Inc
666541IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. The low intrinsic GTPase activity of RAB43 is activated by USP6NL. Involved in retrograde transport from the endocytic pathway to the Golgi apparatus. Involved in the transport of Shiga toxin from early and recycling endosomes to the trans-Golgi network. Required for the structural integrity of the Golgi complex. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.Specifications
| RAB43 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ISY1, RAB11B, RAB41, RAB43, Ras related protein Rab 41, Ras related protein Rab 43 | |
| RAB43 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3B2A8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q86YS6 | |
| 339122 | |
| RAB43 Fusion Protein Ag26911 | |
| Primary | |
| -20°C | |
| RAB43 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title