Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab5a Antibody, Novus Biologicals™
SDP

Catalog No. NB018406 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018406 0.025 mL
NBP120255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018406 Supplier Novus Biologicals Supplier No. NBP1202550.025ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Rab5a Polyclonal specifically detects Rab5a in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rab5a
Applications Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1: 300- 1: 2000, Immunocytochemistry/Immunofluorescence
Formulation Whole antisera with No Preservative
Gene Accession No. Q9CQD1
Gene Alias RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A
Gene Symbols RAB5A
Host Species Sheep
Immunogen A synthetic peptide from the internal region of mouse Ras-related protein Rab-5A (RAB5A) conjugated to an immunogenic carrier protein has been used as the immunogen. The antigen is homologous in rat.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Endosome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5868
Test Specificity RAB5A.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.