Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab5a Antibody, Novus Biologicals™

Sheep Polyclonal Antibody
$206.50 - $478.00
Specifications
| Antigen | Rab5a |
|---|---|
| Concentration | LYOPH |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Antisera |
Description
Rab5a Polyclonal specifically detects Rab5a in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Rab5a | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse, Rat | |
| RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A | |
| RAB5A | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Unconjugated | |
| Sheep | |
| Endosome Markers, Signal Transduction | |
| Q9CQD1 | |
| 5868 | |
| IgG | |
| Unpurified | |
| RAB5A. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title