Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB6A Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB100971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100971 Supplier Novus Biologicals Supplier No. NBP297111R

Rabbit Polyclonal Antibody

RAB6A Polyclonal specifically detects RAB6A in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RAB6A
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias GTP-binding protein RAB6A', GTP-binding protein RAB6B, Oncogene RAB6, Rab GTPase, Rab-6, RAB6, member RAS oncogene family, RAB6A', Rab6A variant 3, RAB6A, member RAS oncogene family, RAB6B, RAB6rab-6, ras-related protein Rab-6A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RAB6A (P20340-2; Met1-Cys208).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5870
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.