Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB6B Antibody (6D12G9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61824 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61824 Supplier Novus Biologicals Supplier No. NBP261824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB6B Monoclonal specifically detects RAB6B in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RAB6B
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6D12G9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias RAB6B, member RAS oncogene family, ras-related protein Rab-6B, small GTPase RAB6B, small GTP-binding protein
Gene Symbols Rab6b
Host Species Mouse
Immunogen Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. Coli.
Molecular Weight of Antigen 23.5 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51560
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.