Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab7a Antibody (10E4), PE, Novus Biologicals™
SDP

Catalog No. NB162862 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162862 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162862 Supplier Novus Biologicals Supplier No. NBP260237PE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rab7a Monoclonal antibody specifically detects Rab7a in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Rab7a
Applications Flow Cytometry
Classification Monoclonal
Clone 100000
Conjugate PE
Formulation PBS
Gene Alias CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a
Host Species Mouse
Immunogen Recombinant human RAB7A (1-207aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.