Learn More
Description
Specifications
Specifications
| Antigen | Rab7a |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Rab7a (NP_076440.1; Met1-Cys207). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
