Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab7a Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB101257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101257 Supplier Novus Biologicals Supplier No. NBP297183C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rab7a Polyclonal specifically detects Rab7a in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Rab7a
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Rab7a fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.