Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB8A Antibody (7G8B11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252548 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252548 Supplier Novus Biologicals Supplier No. NBP252548

Mouse Monoclonal Antibody

RAB8A Monoclonal specifically detects RAB8A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen RAB8A
Applications Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
Classification Monoclonal
Clone 7G8B11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias mel transforming oncogene (derived from cell line NK14), mel transforming oncogene (derived from cell line NK14)- RAB8 homolog, MELras-associated protein RAB8, Oncogene c-mel, RAB8A, member RAS oncogene family, RAB8mel transforming oncogene (RAB8 homolog), ras-related protein Rab-8A
Gene Symbols RAB8A
Host Species Mouse
Immunogen Purified recombinant fragment of human Rab8 (AA: 68-207) expressed in E. Coli.
Molecular Weight of Antigen 23.7 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4218
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.