Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB8A Antibody, Novus Biologicals™
SDP

Catalog No. NB094347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB094347 Supplier Novus Biologicals Supplier No. NBP304519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB8A Polyclonal specifically detects RAB8A in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RAB8A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3. with 0.05% Proclin 300
Gene Alias mel transforming oncogene (derived from cell line NK14), mel transforming oncogene (derived from cell line NK14)- RAB8 homolog, MELras-associated protein RAB8, Oncogene c-mel, RAB8A, member RAS oncogene family, RAB8mel transforming oncogene (RAB8 homolog), ras-related protein Rab-8A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human RAB8A (NP_005361.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4218
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.