Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB8A Antibody, Novus Biologicals™
SDP

Catalog No. NB10093433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093433 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093433 Supplier Novus Biologicals Supplier No. NB10093433

Goat Polyclonal Antibody

RAB8A Polyclonal specifically detects RAB8A in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen RAB8A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005361.2
Gene Alias mel transforming oncogene (derived from cell line NK14), mel transforming oncogene (derived from cell line NK14)- RAB8 homolog, MELras-associated protein RAB8, Oncogene c-mel, RAB8A, member RAS oncogene family, RAB8mel transforming oncogene (RAB8 homolog), ras-related protein Rab-8A
Gene Symbols RAB8A
Host Species Goat
Immunogen Peptide with sequence C-NQGVKITPDQQKR corresponding to C-Terminus according to NP_005361.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4218
Test Specificity Cross reacts with Human. Expected from sequence similarity: Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.