Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB9A Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB325555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325555 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325555 Supplier Novus Biologicals Supplier No. NBP299423JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB9A Polyclonal antibody specifically detects RAB9A in Mouse samples. It is validated for Western Blot

Specifications

Antigen RAB9A
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB9A
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9367
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.