Learn More
Specifications
Specifications
| Antigen | Clusterin/APOJ |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 171% glycerol. |
| Gene Accession No. | P10909 |
| Host Species | Rabbit |
| Immunogen | The antiserum was produced against synthesized peptide derived from human CLU. The immunogen region is: C-terminal. |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
