Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-EDG3, Polyclonal,

Catalog No. ABC501MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC501MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC501MI Supplier MilliporeSigma™ Supplier No. ABC501
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects EDG3 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

EDG3, also known as Sphingosine 1-phosphate receptor 3, S1P receptor 3 or S1P3 or Endothelial differentiation G-protein coupled receptor 3 or Sphingosine 1-phosphate receptor Edg-3 or S1P receptor Edg-3, and encoded by the gene name S1PR3 or EDG3, EDG3 is a member of the EDG family of receptors, which are G protein-coupled receptors. It has been identified as a functional receptor for sphingosine 1-phosphate (S1P) and likely contributes to the regulation of angiogenesis and vascular endothelial cell function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q99500
Gene Symbols S1PR3; EDG3
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the C-terminus of human EDG3.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_005217
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.