Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Fas/TNFRSF6/CD95, Azide Free, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP23311802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23311802 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23311802 Supplier Novus Biologicals Supplier No. NBP2331180.2MG

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml
Formulation 10mM PBS - BSA free with No Preservative
Gene Alias Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Gene Symbols FAS
Host Species Rabbit
Immunogen Amino acids KDITSDSENSNFRNEIQSLV of human FAS (CD95) were used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Diabetes Research, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity There are multiple isoforms of this protein. Researchers are encouraged to use an online tool like NCBI BLAST to determine if the antibody is suitable for their use.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.