Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Luciferase, FITC, Polyclonal Antibody, Novus Biologicals™
SDP

Catalog No. NB100202301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100202301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100202301 Supplier Novus Biologicals Supplier No. NB10020230.1MG

Rabbit Polyclonal Antibody

Luciferase Polyclonal specifically detects Luciferase in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Product Image.

Specifications

Antigen Luciferase
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Western Blot 1:500-1:5000, ELISA 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:100
Formulation PBS with 0.05% Sodium Azide
Gene Alias ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
Host Species Rabbit
Immunogen Luciferase from Photobacterium Vibrio fischerii.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Test Specificity bacterial Luciferase
Target Species Bacteria, Firefly (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.