Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Trypsin 1/PRSS1, Clone: A16-N, Novus Biologicals™
SDP

Catalog No. NBP13013505 Shop All R&D Systems Products
Change view
Click to view available options
0.05mL; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP13013505 0.05mL; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP13013505 Supplier Novus Biologicals Supplier No. NBP1301350.05ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Trypsin 1/PRSS1 Monoclonal specifically detects Trypsin 1/PRSS1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Trypsin 1/PRSS1
Applications Western Blot, ELISA
Classification Monoclonal
Clone A16-N
Concentration 2.7mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:4000, ELISA 1:20000-1:50000
Formulation 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P07477
Gene Alias Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Gene Symbols PRSS1
Host Species Rabbit
Immunogen Peptide derived from near the N terminus of human pancreatic cationic trypsin.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644
Test Specificity This antibody is specific for the N terminus of human pancreatic cationic trypsin.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.