Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabenosyn 5 Antibody, Novus Biologicals™
SDP

Catalog No. NB300813 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300813 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300813 Supplier Novus Biologicals Supplier No. NB300813
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

Rabenosyn 5 Polyclonal specifically detects Rabenosyn 5 in Human samples. It is validated for Western Blot, Peptide ELISA, Knockout Validated.

Specifications

Antigen Rabenosyn 5
Applications Western Blot, ELISA, Immunoassay
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1.5 μg/mL, Peptide ELISA Detection limit 1:64000, Knockout Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_071735.
Gene Alias 110 kDa protein, FLJ34993, FYVE finger-containing Rab5 effector protein rabenosyn-5, FYVE-finger-containing Rab5 effector protein rabenosyn-5, MGC126210, Rabenosyn-5, Zinc finger FYVE domain-containing protein 20, zinc finger, FYVE domain containing 20
Gene Symbols ZFYVE20
Host Species Goat
Immunogen Peptide with sequence C-ELKHTLAKQKGGTD corresponding to C-Terminus according to NP_071735.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64145
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.