Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABEP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB045888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045888 Supplier Novus Biologicals Supplier No. NBP297637H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RABEP1 Polyclonal specifically detects RABEP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen RABEP1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias neurocrescin, rab GTPase-binding effector protein 1, RAB5EPRABPT5A, rabaptin, RAB GTPase binding effector protein 1, rabaptin-4, rabaptin-5, Rabaptin-5alpha, RABPT5rabaptin-5alpha, Renal carcinoma antigen NY-REN-17
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RABEP1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9135
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.