Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABEP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP147285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147285 Supplier Novus Biologicals Supplier No. NBP147285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RABEP1 Polyclonal specifically detects RABEP1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen RABEP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias neurocrescin, rab GTPase-binding effector protein 1, RAB5EPRABPT5A, rabaptin, RAB GTPase binding effector protein 1, rabaptin-4, rabaptin-5, Rabaptin-5alpha, RABPT5rabaptin-5alpha, Renal carcinoma antigen NY-REN-17
Gene Symbols RABEP1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human Rabaptin, RAB GTPase Binding Effector Protein 1 using the numbering given in entry NP_004694.2 (GeneID 9135).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9135
Test Specificity Human. Based on 100% sequence identity, this antibody is expeceted to react with Chimpanzee.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.