Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABGGTA Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB047650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047650 Supplier Novus Biologicals Supplier No. NBP297779AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RABGGTA Polyclonal specifically detects RABGGTA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RABGGTA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.60, Geranylgeranyl transferase type II subunit alpha, geranylgeranyl transferase type-2 subunit alpha, protein prenyltransferase alpha subunit repeat containing 3, PTAR3, Rab geranylgeranyltransferase subunit alpha, Rab geranyl-geranyltransferase subunit alpha, Rab geranylgeranyltransferase, alpha subunit, Rab GG transferase alpha, Rab GGTase alpha
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RABGGTA fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5875
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.