Learn More
Description
Specifications
Specifications
| Antigen | RABGGTA |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | EC 2.5.1.60, Geranylgeranyl transferase type II subunit alpha, geranylgeranyl transferase type-2 subunit alpha, protein prenyltransferase alpha subunit repeat containing 3, PTAR3, Rab geranylgeranyltransferase subunit alpha, Rab geranyl-geranyltransferase subunit alpha, Rab geranylgeranyltransferase, alpha subunit, Rab GG transferase alpha, Rab GGTase alpha |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human RABGGTA fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
