Learn More
Description
Specifications
Specifications
| Antigen | Rabies Virus |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1C5 |
| Conjugate | DyLight 405 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50 mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | It has a virus-neutralizing activity in respect to the majority of standard RV and rabies-relative viruses: CVS, Lagos bat, Mokola, Flury lep, Duvenhage and the field isolates |
| Quantity | 0.2 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
