Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabies Virus Antibody (1C5cc), Novus Biologicals™
SDP

Catalog No. NB001366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001366 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001366 Supplier Novus Biologicals Supplier No. NBP2116300.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rabies Virus
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C5
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.4)
Host Species Mouse
Immunogen It has a virus-neutralizing activity in respect to the majority of standard RV and rabies-relative viruses: CVS, Lagos bat, Mokola, Flury lep, Duvenhage and the field isolates
Quantity 0.2 mg
Target Species Virus
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.