Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABL2A Antibody (OTI4A3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N273777J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273777J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273777J646 Supplier Novus Biologicals Supplier No. NBP273777JF646

Mouse Monoclonal Antibody

RABL2A Monoclonal specifically detects RABL2A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RABL2A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4A3
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ78724, MGC117180, RAB, member of RAS oncogene family-like 2A, rab-like protein 2A
Gene Symbols RABL2A
Host Species Mouse
Immunogen Full length recombinant protein of human RABL2A (NP_009013) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11159.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.