Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABL4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP18717025UL
Description
RABL4 Polyclonal specifically detects RABL4 in Human samples. It is validated for Western Blot.Specifications
RABL4 | |
Polyclonal | |
Western Blot 0.4 ug/ml | |
intraflagellar transport 27 homolog (Chlamydomonas), intraflagellar transport protein 27 homolog, member of RAS oncogene family-like 4, Putative GTP-binding protein RAY-like, RABL4, Rab-like protein 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
IFT27 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TNEESFNNCSKWLEKARSQAPGISLPGVLVGNKTDLAGRRAVDSAEARAWALGQGLECFETSVKEMENFEAPFHCLAKQFHQLYREKVEV | |
25 μL | |
Signal Transduction | |
11020 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction