Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABL4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$382.00 - $646.00
Specifications
Antigen | RABL4 |
---|---|
Dilution | Western Blot 0.4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RABL4 Polyclonal specifically detects RABL4 in Human samples. It is validated for Western Blot.Specifications
RABL4 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
intraflagellar transport 27 homolog (Chlamydomonas), intraflagellar transport protein 27 homolog, member of RAS oncogene family-like 4, Putative GTP-binding protein RAY-like, RABL4, Rab-like protein 4 | |
IFT27 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11020 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TNEESFNNCSKWLEKARSQAPGISLPGVLVGNKTDLAGRRAVDSAEARAWALGQGLECFETSVKEMENFEAPFHCLAKQFHQLYREKVEV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title