Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabphilin 3A, p Ser234 Antibody, Novus Biologicals™
SDP

Catalog No. NB300743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300743 Supplier Novus Biologicals Supplier No. NB300743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabphilin 3A Polyclonal specifically detects Rabphilin 3A in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Rabphilin 3A (p Ser234)
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:400
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P47709
Gene Alias Exophilin-1, KIAA0985exophilin-1, rabphilin, rabphilin 3A homolog (mouse), rabphilin-3A
Gene Symbols RPH3A
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser234 conjugated to KLH. Accession # P47709
Molecular Weight of Antigen 82 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 22895
Test Specificity Specific for the ∼82k rabphilin protein phosphorylated at Ser234. The immunolabeling of the rabphilin band was completely blocked by the phosphopeptide used as antigen while the dephosphopeptide had no effect on the immunolabeling.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.