Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABV Glycoprotein, serum free Mouse anti-Virus, Clone: D1-25, MilliporeSigma™

Mouse Monoclonal Antibody

$198.30 - $503.50

Specifications

Antigen RABV Glycoprotein, serum free
Clone D1-25
Applications ELISA, Immunoprecipitation, Neutralization
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF196725
View Documents
MilliporeSigma
MABF196725UG
25 μg
Each for $198.30
Only null left
Add to Cart
 
MABF1967100
View Documents
MilliporeSigma
MABF1967100UG
100 μg
Each for $503.50
Only null left
Add to Cart
 
Description

Description

RABV Glycoprotein, serum free Monoclonal antibody specifically detects RABV Glycoprotein, serum free in Virus samples. It is validated for ELISA, Immunoprecipitation, Neutralizing.
Specifications

Specifications

RABV Glycoprotein, serum free
ELISA, Immunoprecipitation, Neutralization
Unconjugated
Mouse
Virus
Inactivated and purified Rabies Virus (Pasteur virus strain).
Primary
Stable for 1 year at 2-8°C from date of receipt.
D1-25
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without azide.
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.