Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABX5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987595
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7595 Supplier Bioss Supplier No. BS6916R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.

Specifications

Antigen RABX5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RABGEF1
Gene Accession No. Q9UJ41
Gene Alias BOS_22847; BTB/POZ domain-containing protein KCTD7; LOC479706; RAB guanine nucleotide exchange factor (GEF) 1; RAB guanine nucleotide exchange factor 1; Rab5 exchange factor; Rab5 GDP/GTP exchange factor; Rab5 guanine nucleotide exchange factor Rabex-5; Rab5ef; Rabaptin-5-associated exchange factor for Rab5; Rabex5; Rabex-5; Rabgef1; RAP1; Ras negative regulator Rabex-5; Ras negative regulator Rabex-5/Rin2; Rin2; zgc:65944
Gene Symbols RABGEF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RABX5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27342
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.