Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1,2,3 Mouse anti-Human, Clone: AT2G10, Novus Biologicals™
SDP

Catalog No. NB036457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036457 50 μg
NB036456 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB036457 Supplier Novus Biologicals Supplier No. NBP28883950UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rac1,2,3 Monoclonal specifically detects Rac1,2,3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Rac1,2,3
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone AT2G10
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 2-5ug, ELISA
Formulation PBS (pH 7.4), 10% Glycerol
Host Species Mouse
Immunogen Recombinant human RAC2 (1-189aa) protein purified from E. coli
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.